Product Details

SNP ID
rs3176597
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:47966498 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCAAGTTCAAAGACTGATGTTGT[A/G]CAGCGCAGGGGCCCCCCACACCCTG
Phenotype
MIM: 603760 MIM: 609721
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HUS1 PubMed Links

Gene Details

Gene
HUS1
Gene Name
HUS1 checkpoint clamp component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004507.3 Intron NP_004498.1
XM_017012115.1 Intron XP_016867604.1
Gene
PKD1L1
Gene Name
polycystin 1 like 1, transient receptor potential channel interacting
There are no transcripts associated with this gene.

View Full Product Details