Product Details

SNP ID
rs12104303
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52458918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAAATTACAGATGTTTGTCATAA[A/G]AGGTTCTTACATATTTGGAAAACCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF534 PubMed Links

Gene Details

Gene
ZNF534
Gene Name
zinc finger protein 534
There are no transcripts associated with this gene.

Gene
ZNF578
Gene Name
zinc finger protein 578
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099694.1 Intron NP_001093164.1
XM_017026302.1 Intron XP_016881791.1

View Full Product Details