Product Details

SNP ID
rs13343310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51930214 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACTCTTATTTCATTTCATGTAAA[C/T]GCTACAATATAATATGTGATCTTTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF613 PubMed Links

Gene Details

Gene
ZNF613
Gene Name
zinc finger protein 613
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031721.3 Intron NP_001026891.2
NM_024840.3 Intron NP_079116.2
XM_005259269.3 Intron XP_005259326.1
XM_011527333.2 Intron XP_011525635.1
XM_017027315.1 Intron XP_016882804.1

View Full Product Details