Product Details

SNP ID
rs12793537
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2883469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTTACACTTGTCAATAACCAGAT[G/T]TGGGAGATGGAGAGTGCCTTTGGCA
Phenotype
MIM: 600856 MIM: 603240
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CDKN1C PubMed Links

Gene Details

Gene
CDKN1C
Gene Name
cyclin dependent kinase inhibitor 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000076.2 1536 UTR 3 NP_000067.1
NM_001122630.1 1536 UTR 3 NP_001116102.1
NM_001122631.1 1536 UTR 3 NP_001116103.1
XM_005252732.4 1536 UTR 3 XP_005252789.1
XM_017017088.1 1536 UTR 3 XP_016872577.1
Gene
SLC22A18AS
Gene Name
solute carrier family 22 member 18 antisense
There are no transcripts associated with this gene.

View Full Product Details