Product Details

SNP ID
rs12861323
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103828300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATAGGCTTTATCCTCCTTCTTGG[G/T]TTATATACACTCTACAGAATTCTCA
Phenotype
MIM: 300285
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RAB9B PubMed Links

Gene Details

Gene
RAB9B
Gene Name
RAB9B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016370.2 Intron NP_057454.1

View Full Product Details