Product Details

SNP ID
rs11001178
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:74842047 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTGTATTATTTTCAGTTTTCCAA[C/T]TTGTGGGTTAGAGGGTATCATAGAC
Phenotype
MIM: 605880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KAT6B PubMed Links

Gene Details

Gene
KAT6B
Gene Name
lysine acetyltransferase 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256468.1 Intron NP_001243397.1
NM_001256469.1 Intron NP_001243398.1
NM_012330.3 Intron NP_036462.2
XM_005269664.2 Intron XP_005269721.1
XM_017016000.1 Intron XP_016871489.1
XM_017016001.1 Intron XP_016871490.1
XM_017016002.1 Intron XP_016871491.1
XM_017016003.1 Intron XP_016871492.1
XM_017016004.1 Intron XP_016871493.1
XM_017016005.1 Intron XP_016871494.1
XM_017016006.1 Intron XP_016871495.1
XM_017016007.1 Intron XP_016871496.1
XM_017016008.1 Intron XP_016871497.1
XM_017016009.1 Intron XP_016871498.1

View Full Product Details