Product Details

SNP ID
rs11083761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:45152378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGAAAACGTCCCAGAGGCGCGCGC[A/G]GCCCTCGGGCGCGCCCCCGGTCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NKPD1 PubMed Links

Gene Details

Gene
NKPD1
Gene Name
NTPase, KAP family P-loop domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198478.3 4037 Missense Mutation CGC,TGC R687C NP_940880.3
XM_011526799.2 4037 Missense Mutation CGC,TGC R714C XP_011525101.1
XM_011526802.2 4037 Missense Mutation CGC,TGC R687C XP_011525104.1
XM_011526804.2 4037 Missense Mutation CGC,TGC R568C XP_011525106.1
XM_011526805.2 4037 Missense Mutation CGC,TGC R512C XP_011525107.1
Gene
PPP1R37
Gene Name
protein phosphatase 1 regulatory subunit 37
There are no transcripts associated with this gene.

View Full Product Details