Product Details

SNP ID
rs4369449
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48420586 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGAAGGAGAAAGCATCTCTATCT[C/T]CCAGTCTCTGTGGGAGGATAGGAGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C12orf54 PubMed Links

Gene Details

Gene
C12orf54
Gene Name
chromosome 12 open reading frame 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152319.3 Intron NP_689532.1
XM_005268636.3 Intron XP_005268693.1
XM_011537896.2 Intron XP_011536198.1
XM_017018796.1 Intron XP_016874285.1

View Full Product Details