Product Details

SNP ID
rs12169716
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25212329 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGGCTACATGGCCACACTCAGAGA[A/G]TCCAAGTGAAAGCCCTACAAAGAGG
Phenotype
MIM: 123620 MIM: 123630
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRYBB2 PubMed Links

Gene Details

Gene
CRYBB2
Gene Name
crystallin beta B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000496.2 670 Intron NP_000487.1
XM_006724141.3 670 UTR 5 XP_006724204.1
XM_011529900.2 670 UTR 5 XP_011528202.1
Gene
CRYBB3
Gene Name
crystallin beta B3
There are no transcripts associated with this gene.

View Full Product Details