Product Details

SNP ID
rs11244568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:125576805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCTGGATGAGGAATGGACTGGG[C/T]CCTCACCGGCTCATAGAACTCTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX36 PubMed Links

Gene Details

Gene
TEX36
Gene Name
testis expressed 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128202.2 489 Intron NP_001121674.1
NM_001318133.1 489 Missense Mutation ACC,GCC T112A NP_001305062.1
XM_005269817.4 489 Intron XP_005269874.1
Gene
TEX36-AS1
Gene Name
TEX36 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details