Product Details

SNP ID
rs11662387
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63479608 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAGGTTTCTAGTATGGGGGGTCT[A/G]TCACTCCTGGGGATCCCATAGTGAC
Phenotype
MIM: 154790
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB5 PubMed Links

Gene Details

Gene
SERPINB5
Gene Name
serpin family B member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002639.4 Intron NP_002630.2
XM_006722483.2 Intron XP_006722546.1

View Full Product Details