Product Details

SNP ID
rs12966175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:64088300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATATGACCTTGCTGGCCAAATAA[A/T]TCTTCTGTTTCCATCTCCTTATTTC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LINC00305 PubMed Links

Gene Details

Gene
LINC00305
Gene Name
long intergenic non-protein coding RNA 305
There are no transcripts associated with this gene.

View Full Product Details