Product Details

SNP ID
rs10791779
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106047865 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATCAGCACAATTTGTTGAGCTAGT[A/T]AGACTTTTATGGTTGGGAATGTCAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KBTBD3 PubMed Links

Gene Details

Gene
KBTBD3
Gene Name
kelch repeat and BTB domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152433.3 7246 Intron NP_689646.2
NM_198439.2 7246 Intron NP_940841.1
XM_006718767.3 7246 UTR 3 XP_006718830.1
XM_011542618.2 7246 UTR 3 XP_011540920.1
XM_011542619.2 7246 UTR 3 XP_011540921.1

View Full Product Details