Product Details

SNP ID
rs6589347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112961636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGCTGCAAACTAAGGATCTCAT[C/T]TGGACTTTGTTTTTCCTGGGAACTG
Phenotype
MIM: 116930
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC101928847 PubMed Links

Gene Details

Gene
LOC101928847
Gene Name
uncharacterized LOC101928847
There are no transcripts associated with this gene.

Gene
NCAM1
Gene Name
neural cell adhesion molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000615.6 390 Silent Mutation ATC,ATT I8I NP_000606.3
NM_001076682.3 390 Silent Mutation ATC,ATT I8I NP_001070150.1
NM_001242607.1 390 Silent Mutation ATC,ATT I8I NP_001229536.1
NM_001242608.1 390 Silent Mutation ATC,ATT I8I NP_001229537.1
NM_181351.4 390 Silent Mutation ATC,ATT I8I NP_851996.2

View Full Product Details