Product Details

SNP ID
rs12793033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:28109869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTTTAAAGGAATTGTACATTTG[A/G]TTAAAGCAGAGAGATTTTGACATTA
Phenotype
MIM: 611271
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF18A PubMed Links

Gene Details

Gene
KIF18A
Gene Name
kinesin family member 18A
There are no transcripts associated with this gene.

Gene
METTL15
Gene Name
methyltransferase like 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113528.1 Intron NP_001107000.1
NM_001297775.1 Intron NP_001284704.1
NM_152636.2 Intron NP_689849.2
XM_011519935.2 Intron XP_011518237.1
XM_011519937.2 Intron XP_011518239.1
XM_011519940.2 Intron XP_011518242.1
XM_011519941.2 Intron XP_011518243.1
XM_017017297.1 Intron XP_016872786.1
XM_017017298.1 Intron XP_016872787.1
XM_017017299.1 Intron XP_016872788.1
XM_017017300.1 Intron XP_016872789.1
XM_017017301.1 Intron XP_016872790.1
XM_017017302.1 Intron XP_016872791.1
XM_017017303.1 Intron XP_016872792.1

View Full Product Details