Product Details

SNP ID
rs12151194
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:53049043 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATAAGTAAAGCCTTCTCTCTGTTC[A/C]CCCAAAACTAAAGTCAATCTCAGTA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ERVV-2 PubMed Links

Gene Details

Gene
ERVV-2
Gene Name
endogenous retrovirus group V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191055.1 398 UTR 5 NP_001177984.1

View Full Product Details