Product Details

SNP ID
rs13433132
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37522250 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGGTGTAAAAAGAGATAAATCA[A/G]AAGAAAGCGCTTTGCCCATAAAATC
Phenotype
MIM: 613110 MIM: 603106
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BLCAP PubMed Links

Gene Details

Gene
BLCAP
Gene Name
bladder cancer associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167820.1 121 Intron NP_001161292.1
NM_001167821.1 121 Intron NP_001161293.1
NM_001167822.2 121 Intron NP_001161294.1
NM_001167823.1 121 Intron NP_001161295.1
NM_001317074.1 121 Intron NP_001304003.1
NM_001317075.1 121 Intron NP_001304004.1
NM_006698.3 121 Intron NP_006689.1
XM_017027599.1 121 UTR 5 XP_016883088.1
Gene
NNAT
Gene Name
neuronatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322802.1 121 Intron NP_001309731.1
NM_005386.3 121 Intron NP_005377.1
NM_181689.2 121 Intron NP_859017.1

View Full Product Details