Product Details

SNP ID
rs12389918
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:143026480 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTTCTCACGGCCAGAACTCCCG[C/T]GGAATGTTTCACTTTGACCAAGGTC
Phenotype
MIM: 300667
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SPANXN4 PubMed Links

Gene Details

Gene
SPANXN4
Gene Name
SPANX family member N4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009613.2 Intron NP_001009613.1
XM_017029543.1 Intron XP_016885032.1
XM_017029544.1 Intron XP_016885033.1

View Full Product Details