Product Details

SNP ID
rs10887933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80464011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCTCATGCAGCCTGTGCCCCTA[A/G]AAGTGTGAGGAGTCAGCCAGAGCGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN14 PubMed Links

Gene Details

Gene
TSPAN14
Gene Name
tetraspanin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128309.1 Intron NP_001121781.1
NM_030927.2 Intron NP_112189.2
XM_006717992.3 Intron XP_006718055.1
XM_011540219.2 Intron XP_011538521.1
XM_011540220.2 Intron XP_011538522.1
XM_011540221.2 Intron XP_011538523.1
XM_011540222.2 Intron XP_011538524.1
XM_017016707.1 Intron XP_016872196.1
XM_017016708.1 Intron XP_016872197.1
XM_017016709.1 Intron XP_016872198.1
XM_017016710.1 Intron XP_016872199.1
XM_017016711.1 Intron XP_016872200.1
XM_017016712.1 Intron XP_016872201.1
XM_017016713.1 Intron XP_016872202.1

View Full Product Details