Product Details

SNP ID
rs11620905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93068332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCTCAAATGACCACATGGAAAC[A/C]CTGTGTGCAGAGCCAGCCCCCCAAG
Phenotype
MIM: 601838
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ITPK1 PubMed Links

Gene Details

Gene
ITPK1
Gene Name
inositol-tetrakisphosphate 1-kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017021262.1 Intron XP_016876751.1
XM_017021263.1 Intron XP_016876752.1
XM_017021264.1 Intron XP_016876753.1
Gene
ITPK1-AS1
Gene Name
ITPK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details