Product Details

SNP ID
rs12217793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:16518714 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTTTACACATTTGCTGATAAAA[C/T]TGGAATCATTTCATTTTAATTGATA
Phenotype
MIM: 615227 MIM: 604446
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1QL3 PubMed Links

Gene Details

Gene
C1QL3
Gene Name
complement C1q like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010908.1 Intron NP_001010908.1
Gene
PTER
Gene Name
phosphotriesterase related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001484.2 Intron NP_001001484.1
NM_001261836.1 Intron NP_001248765.1
NM_001261837.1 Intron NP_001248766.1
NM_001261838.1 Intron NP_001248767.1
NM_030664.4 Intron NP_109589.2
XM_017016927.1 Intron XP_016872416.1
XM_017016928.1 Intron XP_016872417.1
XM_017016929.1 Intron XP_016872418.1
XM_017016930.1 Intron XP_016872419.1

View Full Product Details