Product Details

SNP ID
rs11061927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:1696301 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCAGTTTCTACTTGTCAACAGATT[C/G]ACCAGCAGCACCCTCCTCCTCACTG
Phenotype
MIM: 607946
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADIPOR2 PubMed Links

Gene Details

Gene
ADIPOR2
Gene Name
adiponectin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024551.2 Intron NP_078827.2
XM_005253789.1 Intron XP_005253846.1
XM_006719018.2 Intron XP_006719081.1
XM_011521024.2 Intron XP_011519326.1

View Full Product Details