Product Details

SNP ID
rs10754209
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:197042447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAATCTGAAAAACAGGAAGCA[A/T]TTCGAGGTATCAGAAAACCTCCAAG
Phenotype
MIM: 134580
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
F13B PubMed Links

Gene Details

Gene
F13B
Gene Name
coagulation factor XIII B chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001994.2 Intron NP_001985.2
XM_011509283.2 Intron XP_011507585.1
XM_011509284.2 Intron XP_011507586.1
XM_011509286.2 Intron XP_011507588.1

View Full Product Details