Product Details

SNP ID
rs11065235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120734463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACGTGTGGGAGACTCGGAGATGA[C/T]GCTCCGCCCCGCACTTAGAGTCTTA
Phenotype
MIM: 606885
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACADS PubMed Links

Gene Details

Gene
ACADS
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000017.3 Intron NP_000008.1
NM_001302554.1 Intron NP_001289483.1

View Full Product Details