Product Details

SNP ID
rs11872317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46936125 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAAAATCCACTTTTCTATGGT[A/G]GAACATTTTACCATACCTATCTCAG
Phenotype
MIM: 614697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KATNAL2 PubMed Links

Gene Details

Gene
KATNAL2
Gene Name
katanin catalytic subunit A1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031303.2 Intron NP_112593.2
XM_005258357.4 Intron XP_005258414.1
XM_005258358.4 Intron XP_005258415.1
XM_005258361.3 Intron XP_005258418.1
XM_006722554.3 Intron XP_006722617.1
XM_011526219.2 Intron XP_011524521.1
XM_011526220.1 Intron XP_011524522.1
XM_011526221.2 Intron XP_011524523.1
XM_011526223.2 Intron XP_011524525.1
XM_017026028.1 Intron XP_016881517.1
XM_017026029.1 Intron XP_016881518.1
XM_017026030.1 Intron XP_016881519.1
XM_017026031.1 Intron XP_016881520.1
XM_017026032.1 Intron XP_016881521.1

View Full Product Details