Product Details

SNP ID
rs13104730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:174490205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCTAACTTCAGTTTAATTATT[A/T]TCTCAATCAGTATACACCAGAGATG
Phenotype
MIM: 601688
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
HPGD PubMed Links

Gene Details

Gene
HPGD
Gene Name
hydroxyprostaglandin dehydrogenase 15-(NAD)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000860.5 2729 UTR 3 NP_000851.2
NM_001145816.2 2729 UTR 3 NP_001139288.1
NM_001256301.1 2729 UTR 3 NP_001243230.1
NM_001256305.1 2729 UTR 3 NP_001243234.1
NM_001256306.1 2729 UTR 3 NP_001243235.1
NM_001256307.1 2729 UTR 3 NP_001243236.1
XM_011531907.1 2729 Intron XP_011530209.1

View Full Product Details