Product Details

SNP ID
rs12514184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:119087583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTTTTGTGACCTCAGATATGTCT[A/C]TTCTTGGGAATAGTCTATGTGCTGA
Phenotype
MIM: 605671
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DMXL1 PubMed Links

Gene Details

Gene
DMXL1
Gene Name
Dmx like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290321.2 Intron NP_001277250.1
NM_001290322.2 Intron NP_001277251.1
NM_005509.5 Intron NP_005500.4
XM_005271909.4 Intron XP_005271966.1
XM_005271910.4 Intron XP_005271967.1
XM_005271912.2 Intron XP_005271969.1
XM_011543212.2 Intron XP_011541514.1
XM_011543213.2 Intron XP_011541515.1
XM_011543214.2 Intron XP_011541516.1
XM_011543215.2 Intron XP_011541517.1
XM_017009141.1 Intron XP_016864630.1
XM_017009142.1 Intron XP_016864631.1
XM_017009143.1 Intron XP_016864632.1
XM_017009144.1 Intron XP_016864633.1
XM_017009145.1 Intron XP_016864634.1
XM_017009146.1 Intron XP_016864635.1
XM_017009147.1 Intron XP_016864636.1
XM_017009148.1 Intron XP_016864637.1

View Full Product Details