Product Details

SNP ID
rs12106305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:38056364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAAATACGCCATGGAAACTCCA[C/T]GGGATCTGAGTGTAGGTTGATGTAC
Phenotype
MIM: 604829
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DSCR4 PubMed Links

Gene Details

Gene
DSCR4
Gene Name
Down syndrome critical region 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005867.3 Intron NP_005858.1

View Full Product Details