Product Details

SNP ID
rs4971088
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:155170407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTTTTTTACCAGCTCTGCCCCA[A/T]ATCTCACATAACTTCCCTACCCTGG
Phenotype
MIM: 600986
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KRTCAP2 PubMed Links

Gene Details

Gene
KRTCAP2
Gene Name
keratinocyte associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173852.3 Intron NP_776251.1
Gene
TRIM46
Gene Name
tripartite motif containing 46
There are no transcripts associated with this gene.

View Full Product Details