Product Details
- SNP ID
-
rs13177669
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:35048497 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACCAAATAGCTAGGTAAGGTTCTG[C/G]ACAGCTGTGGATCATCTTATGTGTC
- Phenotype
-
MIM: 612471
MIM: 176761
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AGXT2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74506298] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGXT2
- Gene Name
- alanine--glyoxylate aminotransferase 2
- Gene
- PRLR
- Gene Name
- prolactin receptor
View Full Product Details