Product Details

SNP ID
rs11466694
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:1204036 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCGCACCAGGCTGTGACAGACCT[C/T]GGGGCCACATGCACTGACTCCTCAG
Phenotype
MIM: 603905 MIM: 600315
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TNFRSF18 PubMed Links

Gene Details

Gene
TNFRSF18
Gene Name
TNF receptor superfamily member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004195.2 737 Missense Mutation CAA,CGA Q200R NP_004186.1
NM_148901.1 737 Intron NP_683699.1
NM_148902.1 737 Intron NP_683700.1
XM_017002722.1 737 Missense Mutation CAA,CGA Q200R XP_016858211.1
Gene
TNFRSF4
Gene Name
TNF receptor superfamily member 4
There are no transcripts associated with this gene.

Gene
TTLL10
Gene Name
tubulin tyrosine ligase like 10
There are no transcripts associated with this gene.

View Full Product Details