Product Details

SNP ID
rs12125479
Assay Type
Functionally tested
NCBI dbSNP Submissions
28
Location
Chr.1:110113389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCCCAGGCTGAAAGCAGCCCTG[A/G]CCCAGGGTCTCCATGGTTCTAGGCA
Phenotype
MIM: 611127
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC01397 PubMed Links

Gene Details

Gene
LINC01397
Gene Name
long intergenic non-protein coding RNA 1397
There are no transcripts associated with this gene.

Gene
UBL4B
Gene Name
ubiquitin like 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203412.1 950 UTR 3 NP_981957.1

View Full Product Details