Product Details
- SNP ID
-
rs12077739
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:111775214 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTGTTGTGCGTGGCAGTGATGAA[C/G]AAGGCTTTACTGATGTGTTTCAGGA
- Phenotype
-
MIM: 606168
MIM: 605411
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DDX20
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114854284] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DDX20
- Gene Name
- DEAD-box helicase 20
There are no transcripts associated with this gene.
- Gene
- KCND3
- Gene Name
- potassium voltage-gated channel subfamily D member 3
View Full Product Details