Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001005376.2 | Intron | NP_001005376.1 | |||
NM_001005377.2 | Intron | NP_001005377.1 | |||
NM_001301037.1 | Intron | NP_001287966.1 | |||
NM_002659.3 | Intron | NP_002650.1 | |||
XM_005258990.4 | Intron | XP_005259047.1 | |||
XM_011527027.2 | Intron | XP_011525329.1 | |||
XM_011527028.2 | Intron | XP_011525330.1 | |||
XM_011527029.1 | Intron | XP_011525331.1 | |||
XM_011527030.2 | Intron | XP_011525332.1 | |||
XM_011527031.2 | Intron | XP_011525333.1 | |||
XM_017026872.1 | Intron | XP_016882361.1 | |||
XM_017026873.1 | Intron | XP_016882362.1 |