Product Details

SNP ID
rs11716295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50232733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTTGGCTGCCATGTGGAGGATA[A/C]ATTGTAGGGAAACAGGAGAGCCAAG
Phenotype
MIM: 139360
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GNAI2 PubMed Links

Gene Details

Gene
GNAI2
Gene Name
G protein subunit alpha i2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166425.1 Intron NP_001159897.1
NM_001282617.1 Intron NP_001269546.1
NM_001282618.1 Intron NP_001269547.1
NM_001282619.1 Intron NP_001269548.1
NM_001282620.1 Intron NP_001269549.1
NM_002070.3 Intron NP_002061.1
Gene
MIR5787
Gene Name
microRNA 5787
There are no transcripts associated with this gene.

View Full Product Details