Product Details

SNP ID
rs12275912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117211630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTCACTTTCTTGTCCTTTCACA[C/G]CAATACTTGGCAAAAGTCAAACAAT
Phenotype
MIM: 604872 MIM: 600818
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100652768 PubMed Links

Gene Details

Gene
LOC100652768
Gene Name
uncharacterized LOC100652768
There are no transcripts associated with this gene.

Gene
PCSK7
Gene Name
proprotein convertase subtilisin/kexin type 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004716.3 Intron NP_004707.2
XM_006718940.3 Intron XP_006719003.1
XM_017018546.1 Intron XP_016874035.1
Gene
TAGLN
Gene Name
transgelin
There are no transcripts associated with this gene.

View Full Product Details