Product Details

SNP ID
rs7257994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:54334624 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTTTTTGAAGTATGAACCTAGAT[C/T]GGAAGGTGAAAAGCAAATATTTAAA
Phenotype
MIM: 607517
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LILRA4 PubMed Links

Gene Details

Gene
LILRA4
Gene Name
leukocyte immunoglobulin like receptor A4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012276.4 Intron NP_036408.4

View Full Product Details