Product Details
- SNP ID
-
rs11235971
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:74002515 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCTGTAAAATGAGGAAAATAACAG[C/T]ATCTGCCTTGTGGAGTTAGTCGTGC
- Phenotype
-
MIM: 602044
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UCP3
PubMed Links
Gene Details
- Gene
- UCP3
- Gene Name
- uncoupling protein 3
View Full Product Details