Product Details

SNP ID
rs10888274
Assay Type
Functionally tested
NCBI dbSNP Submissions
52
Location
Chr.1:247916661 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAGGATTAGCCACTTCAGTAAGAT[A/C]CAACATGCAAAGGCAGAGGTGTCAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR2T8 PubMed Links

Gene Details

Gene
OR2T8
Gene Name
olfactory receptor family 2 subfamily T member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005522.1 2524 Intron NP_001005522.1
XM_011544178.2 2524 Intron XP_011542480.1
XM_017001172.1 2524 UTR 5 XP_016856661.1

View Full Product Details