Product Details

SNP ID
rs12909277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:68288574 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGTGGGAAAGATCAGACTCTAGCA[A/G]TAGTCTAATTTTGTATGTTAAGTAG
Phenotype
MIM: 613539 MIM: 604789
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FEM1B PubMed Links

Gene Details

Gene
FEM1B
Gene Name
fem-1 homolog B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015322.4 Intron NP_056137.1
XM_017021859.1 Intron XP_016877348.1
Gene
ITGA11
Gene Name
integrin subunit alpha 11
There are no transcripts associated with this gene.

View Full Product Details