Product Details

SNP ID
rs13063790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194592579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGAACTCTGACCTGAGAGAGCC[A/C]TGAAAATCCCATTCATCCTCAGTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TMEM44 PubMed Links

Gene Details

Gene
TMEM44
Gene Name
transmembrane protein 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011655.2 Intron NP_001011655.1
NM_001166305.1 Intron NP_001159777.1
NM_001166306.1 Intron NP_001159778.1
NM_138399.4 Intron NP_612408.3
XM_005269371.4 Intron XP_005269428.1
XM_011513318.2 Intron XP_011511620.1
XM_011513319.2 Intron XP_011511621.1
XM_011513320.2 Intron XP_011511622.1
XM_011513321.2 Intron XP_011511623.1
XM_011513322.2 Intron XP_011511624.1
XM_017007517.1 Intron XP_016863006.1
XM_017007518.1 Intron XP_016863007.1
Gene
TMEM44-AS1
Gene Name
TMEM44 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details