Product Details

SNP ID
rs6439363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:132729506 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATGGTTATATTCTGATGGTTATAT[A/G]TCTGTTACATTCTGACGCCAAAAGG
Phenotype
MIM: 608002
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPHP3 PubMed Links

Gene Details

Gene
NPHP3
Gene Name
nephronophthisis 3 (adolescent)
There are no transcripts associated with this gene.

Gene
NPHP3-ACAD11
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
NPHP3-AS1
Gene Name
NPHP3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details