Product Details
- SNP ID
-
rs10953502
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:106251985 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTGCATATGACATTATCTAATATTT[C/T]TGTCATTGTGTTGCTTCAAACCACA
- Phenotype
-
MIM: 608764
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NAMPT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs530135795] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NAMPT
- Gene Name
- nicotinamide phosphoribosyltransferase
View Full Product Details