Product Details

SNP ID
rs6471887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:60519937 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTAATGCAGAGTGTACCCATAGTT[A/G]TAAGATACACTCAGTGTGGCCCTCC
Phenotype
MIM: 179509
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB2A PubMed Links

Gene Details

Gene
RAB2A
Gene Name
RAB2A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242644.1 Intron NP_001229573.1
NM_002865.2 Intron NP_002856.1

View Full Product Details