Product Details

SNP ID
rs11833291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6380867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGTCCTGGGGTGCTCGGTCTGG[A/G]TCTGGTGTTGCCCAGTGGAAATCAG
Phenotype
MIM: 600979 MIM: 600228
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LTBR PubMed Links

Gene Details

Gene
LTBR
Gene Name
lymphotoxin beta receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270987.1 Intron NP_001257916.1
NM_002342.2 Intron NP_002333.1
XM_005253688.1 Intron XP_005253745.1
XM_006718983.2 Intron XP_006719046.1
Gene
SCNN1A
Gene Name
sodium channel epithelial 1 alpha subunit
There are no transcripts associated with this gene.

View Full Product Details