Product Details

SNP ID
rs12304518
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:3813669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTAGTCAGTTCTCTTGCTGGAAG[C/T]AAGGTACAAAGTAAGGACCAGAAAA
Phenotype
MIM: 616706
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARP11 PubMed Links

Gene Details

Gene
PARP11
Gene Name
poly(ADP-ribose) polymerase family member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286521.1 Intron NP_001273450.1
NM_001286522.1 Intron NP_001273451.1
NM_020367.5 Intron NP_065100.2
XM_005253711.4 Intron XP_005253768.1
XM_011520970.1 Intron XP_011519272.1
XM_011520971.2 Intron XP_011519273.1
XM_011520973.2 Intron XP_011519275.1
XM_017019670.1 Intron XP_016875159.1
XM_017019671.1 Intron XP_016875160.1

View Full Product Details