Product Details

SNP ID
rs10779632
Assay Type
Functionally tested
NCBI dbSNP Submissions
60
Location
Chr.1:215005140 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTCCTCTTATTTGACTTTTTTAC[A/G]TGGCTTCTACAATCATAAAACAAGC
Phenotype
MIM: 603219
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNK2 PubMed Links

Gene Details

Gene
KCNK2
Gene Name
potassium two pore domain channel subfamily K member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017424.2 Intron NP_001017424.1
NM_001017425.2 Intron NP_001017425.2
NM_014217.3 Intron NP_055032.1
XM_011509522.2 Intron XP_011507824.1
XM_011509523.1 Intron XP_011507825.1
XM_011509524.2 Intron XP_011507826.1
XM_017001248.1 Intron XP_016856737.1
XM_017001249.1 Intron XP_016856738.1

View Full Product Details