Product Details

SNP ID
rs10784339
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:63144866 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATTTAAAAGGAGAAAATACAACT[C/G]GGTAGGGTGATATGCACTTTTTTTT
Phenotype
MIM: 600821
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
AVPR1A PubMed Links

Gene Details

Gene
AVPR1A
Gene Name
arginine vasopressin receptor 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000706.4 5724 UTR 3 NP_000697.1

View Full Product Details