Product Details

SNP ID
rs12844744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:17745754 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAAATTAAGAGCAGTTTTGTAC[A/T]CACTTTCAGTATAAAACTCTAAACC
Phenotype
MIM: 300457 MIM: 300227
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NHS PubMed Links

Gene Details

Gene
NHS
Gene Name
NHS actin remodeling regulator
There are no transcripts associated with this gene.

Gene
SCML1
Gene Name
sex comb on midleg-like 1 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037535.2 Intron NP_001032624.1
NM_001037536.2 Intron NP_001032625.1
NM_001037540.2 Intron NP_001032629.1
NM_006746.5 Intron NP_006737.2
XM_005274578.2 Intron XP_005274635.1
XM_005274579.2 Intron XP_005274636.1
XM_006724508.3 Intron XP_006724571.1
XM_006724509.3 Intron XP_006724572.1
XM_011545564.2 Intron XP_011543866.1
XM_017029723.1 Intron XP_016885212.1

View Full Product Details