Product Details

SNP ID
rs11636318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:81333044 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAGATGTTTTTTCTTGTAAGAGTCA[C/T]ATTCATTAATGACATAGTATCTGGG
Phenotype
MIM: 607050
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STARD5 PubMed Links

Gene Details

Gene
STARD5
Gene Name
StAR related lipid transfer domain containing 5
There are no transcripts associated with this gene.

Gene
TMC3
Gene Name
transmembrane channel like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080532.1 2678 Missense Mutation TAT,TGT Y893C NP_001074001.1
Gene
TMC3-AS1
Gene Name
TMC3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details